Assay Details
| Gene Symbol: CYBRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193742.2 | NP_001180671.1 | 584 | 79 | ||
| GenBank mRNA | JU323390.1 | - | 481 | 79 | ||
| JU470763.1 | - | 574 | 79 | |||
| JV048284.1 | - | 464 | 79 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CYBRD1
Genomic Map