Assay Details
| Gene Symbol: ITPKC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015123993.2 | XP_014979479.1 | 1402 | 133 | ||
| XM_015123994.2 | XP_014979480.1 | 1402 | 133 | |||
| GenBank mRNA | JU494009.1 | - | 1047 | 133 | ||
| JV662209.1 | - | 1094 | 133 | |||
More Information
Set Membership: |
|

ITPKC
Genomic Map