Assay Details
| Gene Symbol: ZNF436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257465.1 | NP_001244394.1 | 504 | 123 | ||
| XM_028843650.1 | XP_028699483.1 | 2629 | 123 | |||
| GenBank mRNA | JU336521.1 | - | 504 | 123 | ||
| JU477330.1 | - | 534 | 123 | |||
| JV048323.1 | - | 712 | 123 | |||
| JV048324.1 | - | 523 | 123 | |||
| JV662371.1 | - | 703 | 123 | |||
More Information
Set Membership: |
|

ZNF436
Genomic Map