Assay Details
| Gene Symbol: TRIM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266728.2 | NP_001253657.1 | 1030 | 71 | ||
| GenBank mRNA | JU322213.1 | - | 1017 | 71 | ||
| JU471161.1 | - | 1030 | 71 | |||
| JV638636.1 | - | 1039 | 71 | |||
More Information
Set Membership: |
|

TRIM8
Genomic Map