Assay Details
| Gene Symbol: NUDT12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194123.1 | NP_001181052.1 | 1077 | 72 | ||
| XM_015140490.2 | XP_014995976.1 | 881 | 72 | |||
| XM_015140491.2 | XP_014995977.1 | 988 | 72 | |||
| XM_015140492.2 | XP_014995978.1 | 671 | 72 | |||
| GenBank mRNA | JU322488.1 | - | 798 | 72 | ||
| JV043968.1 | - | 836 | 72 | |||
More Information
Set Membership: |
|

NUDT12
Genomic Map