Assay Details
| Gene Symbol: TOR1AIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261130.1 | NP_001248059.1 | 820 | 90 | ||
| GenBank mRNA | JU323370.1 | - | 820 | 90 | ||
| JU474504.1 | - | 820 | 90 | |||
More Information
Set Membership: |
|

TOR1AIP2
Genomic Map