Assay Details
| Gene Symbol: TMEM106A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194813.1 | NP_001181742.1 | 350 | 83 | ||
| XM_001113491.4 | XP_001113491.1 | 614 | 83 | |||
| XM_015119753.2 | XP_014975239.1 | 392 | 83 | |||
| XM_015119754.2 | XP_014975240.1 | 614 | 83 | |||
| XM_028835637.1 | XP_028691470.1 | 484 | 83 | |||
| XM_028835638.1 | XP_028691471.1 | 605 | 83 | |||
| GenBank mRNA | JV641383.1 | - | 325 | 83 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TMEM106A
Genomic Map