Assay Details
| Gene Symbol: DNAI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001108527.4 | XP_001108527.2 | 1289 | 130 | ||
| XM_015144182.2 | XP_014999668.2 | 1171 | 130 | |||
| XM_028831893.1 | XP_028687726.1 | 1243 | 130 | |||
More Information
Set Membership: |
|

DNAI3
Genomic Map