Assay Details
| Gene Symbol: MOB3C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260946.1 | NP_001247875.1 | 824 | 92 | ||
| XM_028843456.1 | XP_028699289.1 | 2431 | 92 | |||
| XM_028843457.1 | XP_028699290.1 | 759 | 92 | |||
| GenBank mRNA | JU319953.1 | - | 824 | 92 | ||
| JU478243.1 | - | 828 | 92 | |||
More Information
Set Membership: |
|

MOB3C
Genomic Map