Assay Details
| Gene Symbol: CHMP4C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193985.1 | NP_001180914.1 | 377 | 70 | ||
| GenBank mRNA | JV665604.1 | - | 163 | 70 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CHMP4C| Gene Symbol: CHMP4C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193985.1 | NP_001180914.1 | 377 | 70 | ||
| GenBank mRNA | JV665604.1 | - | 163 | 70 | ||
Set Membership: |
|