Assay Details
| Gene Symbol: PROCA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015118906.2 | XP_014974392.1 | 717 | 117 | ||
| XM_015118907.2 | XP_014974393.1 | 721 | 117 | |||
| XM_015118908.2 | XP_014974394.1 | 369 | 117 | |||
| XM_015118910.2 | XP_014974396.1 | 693 | 117 | |||
| XM_015118911.2 | XP_014974397.1 | 632 | 117 | |||
| XM_015118913.2 | XP_014974399.1 | 607 | 117 | |||
More Information
Set Membership: |
|

PROCA1
Genomic Map