Assay Details
| Gene Symbol: SH3RF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001099248.4 | XP_001099248.2 | 1396 | 74 | ||
| XM_015141123.2 | XP_014996609.1 | 1396 | 74 | |||
| XM_028849816.1 | XP_028705649.1 | 1396 | 74 | |||
| GenBank mRNA | JV607163.1 | - | 126 | 74 | ||
More Information
Set Membership: |
|

SH3RF2
Genomic Map