Assay Details
| Gene Symbol: CCDC63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015152793.2 | XP_015008279.2 | 1380 | 126 | ||
| XM_015152794.2 | XP_015008280.1 | 966 | 126 | |||
| XM_028829873.1 | XP_028685706.1 | 1166 | 126 | |||
| XM_028829874.1 | XP_028685707.1 | 1171 | 126 | |||
| XM_028829875.1 | XP_028685708.1 | 1171 | 126 | |||
| GenBank mRNA | JU481324.1 | - | 775 | 126 | ||
More Information
Set Membership: |
|

CCDC63
Genomic Map