Assay Details
| Gene Symbol: LACC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193989.2 | NP_001180918.1 | 766 | 90 | ||
| XM_015120901.2 | XP_014976387.1 | 850 | 90 | |||
| XM_028837113.1 | XP_028692946.1 | 842 | 90 | |||
| XM_028837114.1 | XP_028692947.1 | 316 | 90 | |||
| GenBank mRNA | JU323204.1 | - | 751 | 90 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028837114.1 transcripts.
Set Membership: |
|

LACC1
Genomic Map