Assay Details
| Gene Symbol: SETD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194171.2 | NP_001181100.1 | 980 | 137 | ||
| XM_015140051.2 | XP_014995537.1 | 989 | 137 | |||
| XM_015140052.2 | XP_014995538.1 | 989 | 137 | |||
| XM_015140053.2 | XP_014995539.1 | 902 | 137 | |||
| XM_028849231.1 | XP_028705064.1 | 724 | 137 | |||
| XM_028849232.1 | XP_028705065.1 | 751 | 137 | |||
| XM_028849233.1 | XP_028705066.1 | 514 | 137 | |||
| GenBank mRNA | JU332963.1 | - | 647 | 137 | ||
More Information
Set Membership: |
|

SETD9
Genomic Map