Assay Details
| Gene Symbol: PHF13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260753.1 | NP_001247682.1 | 301 | 92 | ||
| GenBank mRNA | JU333080.1 | - | 127 | 92 | ||
| JV044376.1 | - | 301 | 92 | |||
| JV606821.1 | - | 274 | 92 | |||
| JV637222.1 | - | 296 | 92 | |||
More Information
Set Membership: |
|

PHF13
Genomic Map