Assay Details
| Gene Symbol: NDUFAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266244.1 | NP_001253173.1 | 219 | 142 | ||
| GenBank mRNA | JU323411.1 | - | 278 | 142 | ||
| JV045666.1 | - | 219 | 142 | |||
| JV598251.1 | - | 327 | 142 | |||
More Information
Set Membership: |
|

NDUFAF2
Genomic Map