Assay Details
| Gene Symbol: UGT3A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015139865.2 | XP_014995351.1 | 249 | 127 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UGT3A2| Gene Symbol: UGT3A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015139865.2 | XP_014995351.1 | 249 | 127 | ||
Set Membership: |
|