Assay Details
| Gene Symbol: CALHM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194618.2 | NP_001181547.2 | 544 | 154 | ||
| XM_015148024.2 | XP_015003510.2 | 3417 | 154 | |||
| XM_028853524.1 | XP_028709357.1 | 3084 | 154 | |||
| XM_028853525.1 | XP_028709358.1 | 4135 | 154 | |||
| XM_028853526.1 | XP_028709359.1 | 3655 | 154 | |||
| XM_028853527.1 | XP_028709360.1 | 4328 | 154 | |||
| GenBank mRNA | JV605456.1 | - | 94 | 154 | ||
More Information
Set Membership: |
|

CALHM3
Genomic Map