Assay Details
| Gene Symbol: SNX20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261262.1 | NP_001248191.1 | 405 | 103 | ||
| GenBank mRNA | JU474278.1 | - | 405 | 103 | ||
| JU474279.1 | - | 405 | 103 | |||
| JU474280.1 | - | 405 | 103 | |||
| JU474281.1 | - | 405 | 103 | |||
More Information
Set Membership: |
|

SNX20
Genomic Map