Assay Details
| Gene Symbol: GLT6D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001097091.4 | XP_001097091.1 | 596 | 125 | ||
| XM_001097205.4 | XP_001097205.1 | 552 | 125 | |||
| XM_015116385.2 | XP_014971871.1 | 329 | 125 | |||
| XM_028834614.1 | XP_028690447.1 | 597 | 125 | |||
| XM_028834615.1 | XP_028690448.1 | 786 | 125 | |||
| XM_028834617.1 | XP_028690450.1 | 329 | 125 | |||
More Information
Set Membership: |
|

GLT6D1
Genomic Map