Assay Details
| Gene Symbol: C1H1orf52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258201.1 | NP_001245130.1 | 491 | 150 | ||
| GenBank mRNA | JU333079.1 | - | 507 | 150 | ||
| JU472774.1 | - | 491 | 150 | |||
| JV046260.1 | - | 492 | 150 | |||
| JV707717.1 | - | 455 | 150 | |||
More Information
Set Membership: |
|

C1H1orf52
Genomic Map