Assay Details
| Gene Symbol: PRICKLE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260650.1 | NP_001247579.1 | 730 | 82 | ||
| XM_015130445.2 | XP_014985931.1 | 444 | 82 | |||
| XM_015130446.2 | XP_014985932.1 | 633 | 82 | |||
| GenBank mRNA | JU333205.1 | - | 730 | 82 | ||
| JV567187.1 | - | 817 | 82 | |||
More Information
Set Membership: |
|

PRICKLE2
Genomic Map