Assay Details
| Gene Symbol: CHRM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257740.2 | NP_001244669.1 | 170 | 118 | ||
| XM_015126482.2 | XP_014981968.1 | 530 | 118 | |||
| XM_015126487.2 | XP_014981973.1 | 486 | 118 | |||
| XM_015126492.2 | XP_014981978.1 | 277 | 118 | |||
| XM_028843478.1 | XP_028699311.1 | 1308 | 118 | |||
| XM_028843481.1 | XP_028699314.1 | 289 | 118 | |||
| XM_028843485.1 | XP_028699318.1 | 688 | 118 | |||
| XM_028843487.1 | XP_028699320.1 | 764 | 118 | |||
| GenBank mRNA | JU321096.1 | - | 173 | 118 | ||
| JV451866.1 | - | 173 | 118 | |||
| JV567059.1 | - | 133 | 118 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001257740.2, XM_015126482.2, XM_015126487.2, XM_015126492.2, XM_028843478.1, XM_028843481.1, XM_028843485.1, XM_028843487.1 transcripts.
Set Membership: |
|

CHRM3
Genomic Map