Assay Details
| Gene Symbol: ZNF425 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015135224.2 | XP_014990710.2 | 1229 | 88 | ||
| GenBank mRNA | JV707983.1 | - | 42 | 88 | ||
| JV707984.1 | - | 42 | 88 | |||
More Information
Set Membership: |
|

ZNF425
Genomic Map