Assay Details
| Gene Symbol: TMEM200B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133442.2 | XP_014988928.1 | 1501 | 127 | ||
| XM_028837112.1 | XP_028692945.1 | 1440 | 127 | |||
| GenBank mRNA | JV651272.1 | - | 1242 | 126 | ||
| JV714850.1 | - | 1144 | 126 | |||
| Gene Symbol: EPB41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JV054354.1 | - | 2138 | 126 | ||
More Information
Set Membership: |
|

TMEM200B
Genomic Map