Assay Details
| Gene Symbol: F8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028842500.1 | XP_028698333.1 | 310 | 75 | ||
| Gene Symbol: LOC114675039 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028842397.1 | XP_028698230.1 | 296 | 75 | ||
| Gene Symbol: LOC703511 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015128572.2 | XP_014984058.1 | 875 | 75 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028842500.1 transcripts.
Set Membership: |
|

F8
Genomic Map