Assay Details
| Gene Symbol: NANOS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015124149.2 | XP_014979635.1 | 1521 | 104 | ||
| GenBank mRNA | JV650114.1 | - | 711 | 104 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NANOS2| Gene Symbol: NANOS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015124149.2 | XP_014979635.1 | 1521 | 104 | ||
| GenBank mRNA | JV650114.1 | - | 711 | 104 | ||
Set Membership: |
|