Assay Details
| Gene Symbol: LOC100428164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001104545.4 | XP_001104545.3 | 3267 | 118 | ||
| XM_015148387.2 | XP_015003873.2 | 1012 | 118 | |||
| XM_028826970.1 | XP_028682803.1 | 998 | 118 | |||
More Information
Set Membership: |
|

LOC100428164
Genomic Map