Assay Details
| Gene Symbol: SPATA6L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015117770.2 | XP_014973256.1 | 373 | 156 | ||
| XM_015117773.2 | XP_014973259.1 | 254 | 156 | |||
| XM_028835183.1 | XP_028691016.1 | 559 | 156 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015117773.2 transcripts.
Set Membership: |
|

SPATA6L
Genomic Map