Assay Details
| Gene Symbol: FMOD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260677.1 | NP_001247606.1 | 979 | 82 | ||
| XM_015118585.2 | XP_014974071.1 | 1436 | 82 | |||
| XM_015118591.2 | XP_014974077.1 | 1436 | 82 | |||
| GenBank mRNA | JU474893.1 | - | 1112 | 82 | ||
| JV046562.1 | - | 979 | 82 | |||
| JV636058.1 | - | 1118 | 82 | |||
More Information
Set Membership: |
|

FMOD
Genomic Map