Assay Details
| Gene Symbol: SRSF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001190739.1 | NP_001177668.1 | 461 | 144 | ||
| XM_015120341.2 | XP_014975827.1 | 615 | 144 | |||
| GenBank mRNA | CN643650.1 | - | 228 | 144 | ||
| JU319605.1 | - | 525 | 144 | |||
| JU335899.1 | - | 525 | 144 | |||
| JU491507.1 | - | 357 | 144 | |||
| JV044458.1 | - | 535 | 144 | |||
| JV047917.1 | - | 535 | 144 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SRSF2
Genomic Map