Assay Details
| Gene Symbol: TLR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015139351.2 | XP_014994837.2 | 2594 | 153 | ||
| XM_015139352.2 | XP_014994838.2 | 2478 | 153 | |||
| GenBank mRNA | EU204933.1 | - | 2440 | 153 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TLR2
Genomic Map