Assay Details
| Gene Symbol: MED20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015136336.2 | XP_014991822.1 | 551 | 83 | ||
| XM_015136337.2 | XP_014991823.1 | 454 | 83 | |||
| XM_015136338.2 | XP_014991824.1 | 396 | 83 | |||
| GenBank mRNA | JU319910.1 | - | 528 | 83 | ||
| JU473441.1 | - | 528 | 83 | |||
| JV048763.1 | - | 467 | 83 | |||
More Information
Set Membership: |
|

MED20
Genomic Map