Assay Details
| Gene Symbol: NDUFB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266522.1 | NP_001253451.1 | 249 | 113 | ||
| XM_015129909.2 | XP_014985395.1 | 364 | 113 | |||
| GenBank mRNA | JU475702.1 | - | 251 | 113 | ||
| JV047148.1 | - | 249 | 113 | |||
| JV507831.1 | - | 264 | 113 | |||
| JV715753.1 | - | 263 | 113 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NDUFB4
Genomic Map