Assay Details
| Gene Symbol: HAND1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001111722.3 | XP_001111722.1 | 197 | 152 | ||
| XM_015141304.2 | XP_014996790.1 | 205 | 152 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001111722.3, XM_015141304.2 transcripts.
Set Membership: |
|

HAND1
Genomic Map