Assay Details
| Gene Symbol: RRAGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194800.2 | NP_001181729.1 | 1583 | 160 | ||
| GenBank mRNA | JU319587.1 | - | 1464 | 160 | ||
| JU470807.1 | - | 1462 | 160 | |||
| JV045682.1 | - | 1477 | 160 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RRAGA
Genomic Map