Assay Details
| Gene Symbol: BTG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266234.1 | NP_001253163.1 | 223 | 65 | ||
| GenBank mRNA | JU328444.1 | - | 85 | 65 | ||
| JU477143.1 | - | 189 | 65 | |||
| JV048206.1 | - | 223 | 65 | |||
More Information
Set Membership: |
|

BTG2
Genomic Map