Assay Details
| Gene Symbol: ZBTB33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266950.1 | NP_001253879.1 | 136 | 135 | ||
| GenBank mRNA | JU332369.1 | - | 157 | 135 | ||
| JU471679.1 | - | 136 | 135 | |||
| JV602718.1 | - | 235 | 135 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001266950.1 transcripts.
Set Membership: |
|

ZBTB33
Genomic Map