Assay Details
| Gene Symbol: HOXD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015110410.2 | XP_014965896.1 | 182 | 105 | ||
| XM_028830864.1 | XP_028686697.1 | 325 | 105 | |||
| XM_028830865.1 | XP_028686698.1 | 2138 | 105 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015110410.2, XM_028830864.1, XM_028830865.1 transcripts.
Set Membership: |
|

HOXD3
Genomic Map