Assay Details
| Gene Symbol: DCUN1D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257454.1 | NP_001244383.1 | 268 | 108 | ||
| XM_015131496.2 | XP_014986982.1 | 432 | 108 | |||
| XM_015131498.2 | XP_014986984.1 | 763 | 108 | |||
| XM_015131499.2 | XP_014986985.1 | 864 | 108 | |||
| XM_015131500.2 | XP_014986986.1 | 314 | 108 | |||
| XM_015131501.2 | XP_014986987.1 | 763 | 108 | |||
| GenBank mRNA | JU320015.1 | - | 268 | 108 | ||
| JU476060.1 | - | 259 | 108 | |||
More Information
Set Membership: |
|

DCUN1D1
Genomic Map