Assay Details
| Gene Symbol: ATF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266632.2 | NP_001253561.1 | 929 | 138 | ||
| GenBank mRNA | JU322509.1 | - | 564 | 138 | ||
| JU487167.1 | - | 519 | 138 | |||
| JV043307.1 | - | 638 | 138 | |||
| JV047142.1 | - | 515 | 138 | |||
| JV507789.1 | - | 499 | 138 | |||
| JV636171.1 | - | 929 | 138 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ATF4
Genomic Map