Assay Details
| Gene Symbol: CRYZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001265811.1 | NP_001252740.1 | 383 | 116 | ||
| XM_015142315.2 | XP_014997801.1 | 451 | 116 | |||
| XM_015142326.2 | XP_014997812.1 | 369 | 116 | |||
| XM_028840906.1 | XP_028696739.1 | 385 | 116 | |||
| GenBank mRNA | JV565070.1 | - | 383 | 116 | ||
| JV707394.1 | - | 388 | 116 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CRYZ
Genomic Map