Assay Details
| Gene Symbol: RAB13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261291.1 | NP_001248220.1 | 348 | 150 | ||
| XM_028845402.1 | XP_028701235.1 | 503 | 150 | |||
| GenBank mRNA | JU320809.1 | - | 342 | 150 | ||
| JV044701.1 | - | 348 | 150 | |||
| JV639435.1 | - | 375 | 150 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay is in the 5'UTR region of the XM_028845402.1 transcripts.
This assay is in the 5'UTR region of the XM_028845402.1 transcripts.
Set Membership: |
|

RAB13
Genomic Map