Assay Details
| Gene Symbol: LOC699924 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015115204.2 | XP_014970690.1 | 1271 | 63 | ||
| Gene Symbol: LOC699676 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028826129.1 | XP_028681962.1 | 1095 | 63 | ||
More Information
Set Membership: |
|

LOC699924
Genomic Map