Assay Details
| Gene Symbol: RBMX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266848.1 | NP_001253777.1 | 141 | 142 | ||
| GenBank mRNA | JU334945.1 | - | 178 | 142 | ||
| JU475948.1 | - | 181 | 142 | |||
| JV047337.1 | - | 141 | 142 | |||
More Information
Set Membership: |
|

RBMX2
Genomic Map