Assay Details
| Gene Symbol: VPS33A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257848.1 | NP_001244777.1 | 313 | 140 | ||
| GenBank mRNA | JU335981.1 | - | 313 | 140 | ||
| JU476860.1 | - | 356 | 140 | |||
| JV045505.1 | - | 319 | 140 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

VPS33A
Genomic Map