Assay Details
| Gene Symbol: GNG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU334063.1 | - | 47 | 140 | ||
| JU471725.1 | - | 69 | 140 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GNG5| Gene Symbol: GNG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU334063.1 | - | 47 | 140 | ||
| JU471725.1 | - | 69 | 140 | |||
Set Membership: |
|