Assay Details
| Gene Symbol: ATP5MC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JV565031.1 | - | 156 | 117 | ||
| JV669189.1 | - | 116 | 117 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATP5MC2| Gene Symbol: ATP5MC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JV565031.1 | - | 156 | 117 | ||
| JV669189.1 | - | 116 | 117 | |||
Set Membership: |
|