Assay Details
| Gene Symbol: IMPACT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266051.1 | NP_001252980.1 | 2367 | 81 | ||
| XM_015121655.2 | XP_014977141.1 | 2642 | 81 | |||
| XM_028837749.1 | XP_028693582.1 | 2320 | 81 | |||
| GenBank mRNA | JU335330.1 | - | 2336 | 81 | ||
| JU335331.1 | - | 2336 | 81 | |||
| JV047564.1 | - | 2367 | 81 | |||
More Information
Set Membership: |
|

IMPACT
Genomic Map